Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR1-1HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302812.1 | NP_001289741.1 | 4, 5 | 519 | ||
NM_178463.3 | NP_848558.1 | 3, 4 | 900 | |||
GenBank mRNA | AK056985.1 | - | 3, 4 | 916 | ||
BC110595.2 | - | 3, 4 | 575 | 40022808 | ||
DA896197.1 | - | 4, 5 | 516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624914.3 | - | 900 | Chr. - - [-] on Build |