Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153244.1 | NP_694976.1 | 2 | 1345 | ||
GenBank mRNA | BC029034.1 | - | 2 | 1345 | 5194035 | |
BC036401.1 | - | 1 | 362 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624760.1 | - | 1345 | Chr. - - [-] on Build |