Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302813.1 | NP_001289742.1 | 3 | 2524 | ||
NM_001302815.1 | NP_001289744.1 | 4 | 2213 | |||
NM_173644.2 | NP_775915.1 | 4 | 2604 | |||
XM_017027828.1 | XP_016883317.1 | 3 | 11329 | |||
XM_017027829.1 | XP_016883318.1 | 4 | 8991 | |||
XM_017027830.1 | XP_016883319.1 | 3 | 8856 | |||
XM_017027831.1 | XP_016883320.1 | 4 | 10675 | |||
XM_017027832.1 | XP_016883321.1 | 4 | 2728 | |||
GenBank mRNA | AK091179.1 | - | 4 | 2604 | ||
AY358539.1 | - | 1 | 4313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000625080.1 | - | 2604 | Chr. - - [-] on Build |