Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130935.1 | 3 | 972 | |||
GenBank mRNA | AK075315.1 | - | 3 | 972 | ||
BC104448.1 | - | 2 | 840 | |||
BC104449.1 | - | 2 | 840 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446232.1 | - | 2 | 840 | Chr. 11 - 67604053 - 67605964 [-] on Build GRCh38 |