Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 3 | 776 | |||
GenBank mRNA | AK057399.1 | - | 3 | 712 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000470346.2 | - | 3 | 712 | Chr. 2 - 238225120 - 238231613 [-] on Build GRCh38 |