Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033978.1 | 2 | 555 | |||
GenBank mRNA | AK125779.1 | - | 2 | 555 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000485805.1 | - | 2 | 555 | Chr. 3 - 64078361 - 64087363 [-] on Build GRCh38 |