Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526860.2 | XP_011525162.1 | 1 | 52 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000602554.1 | - | 2 | 80 | Chr. 19 - 49331617 - 49340282 [-] on Build GRCh38 |