Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002457.3 | NP_002448.3 | 13512 | |||
GenBank mRNA | AF257167.1 | - | 5 | 933 | ||
L21998.1 | - | 13512 | ||||
M86523.1 | - | 1 | 12 | |||
M94132.1 | - | 5 | 900 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000616479.1 | - | 12 | Chr. - - [-] on Build |