Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100134040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL162052.1 | - | 1 | 955 |
Gene Symbol: LOC101929998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127966.1 | - | 1 | 1536 |
Gene Symbol: LOC107984014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012898.1 | XP_016868387.1 | 5 | 1123 |
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC113028.1 | - | 1 | 126 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425591.1 | - | 1 | 1532 | Chr. 7 - 154055585 - 154059561 [-] on Build GRCh38 |