Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100507175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103861.1 | 3 | 1160 | |||
GenBank mRNA | AK055974.1 | - | 3 | 1160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000400306.2 | - | 3 | 1160 | Chr. 12 - 67520082 - 67567126 [+] on Build GRCh38 |