Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC5OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195576.1 | NP_001182505.1 | 4 | 460 | ||
NM_001195577.1 | NP_001182506.1 | 4 | 441 | |||
NM_001195578.1 | NP_001182507.1 | 4 | 395 | |||
XM_011530838.2 | XP_011529140.1 | 4 | 389 | |||
GenBank mRNA | DB076612.1 | - | 4 | 460 | ||
DB096871.1 | - | 4 | 452 | |||
DB458957.1 | - | 4 | 395 |