Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC5OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195576.1 | NP_001182505.1 | 4 | 2574 | ||
NM_001195577.1 | NP_001182506.1 | 4 | 2555 | |||
NM_001195578.1 | NP_001182507.1 | 4 | 2509 | |||
XM_011530838.2 | XP_011529140.1 | 4 | 2503 |