Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC151121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122040.1 | 4 | 617 | |||
NR_122041.1 | 3 | 529 | ||||
NR_122042.1 | 3 | 540 | ||||
GenBank mRNA | AK056598.1 | - | 4 | 574 | ||
DA821832.1 | - | 3 | 378 | |||
DA824517.1 | - | 3 | 353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000375987.3 | - | 574 | Chr. 2 - 129242388 - 129242693 [-] on Build GRCh38 |