Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103538.1 | 1 | 501 | |||
GenBank mRNA | BX118328.1 | - | 1 | 501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000377186.3 | - | 2 | 213 | Chr. 6 - 28943877 - 28944537 [+] on Build GRCh38 |