Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024607.2 | NP_001019778.1 | 1 | 114 | ||
GenBank mRNA | BG211856.1 | - | 1 | 115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624695.1 | - | 114 | Chr. - - [-] on Build |