Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034140.1 | 2 | 837 | |||
GenBank mRNA | AK123808.1 | - | 2 | 843 | ||
BC028195.1 | - | 2 | 837 | 5163453 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000538094.1 | - | 2 | 837 | Chr. 12 - 9055586 - 9065070 [-] on Build GRCh38 |