Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 4, 5 | 477 | |||
NR_027791.3 | 3, 4 | 395 | ||||
NR_111004.2 | 4, 5 | 522 | ||||
NR_111005.2 | 4, 5 | 522 | ||||
NR_136541.1 | 5, 6 | 602 | ||||
NR_136542.1 | 5, 6 | 602 | ||||
NR_136543.1 | 3, 4 | 395 | ||||
NR_136551.1 | 3, 4 | 430 | ||||
NR_136552.1 | 3, 4 | 430 | ||||
GenBank mRNA | AF486622.1 | - | 4, 5 | 403 | ||
AK095614.1 | - | 4, 5 | 477 | |||
AL700268.1 | - | 3, 4 | 306 | |||
BC130602.1 | - | 3, 4 | 379 | |||
BC130606.1 | - | 3, 4 | 379 | |||
CF135136.1 | - | 3, 4 | 402 | |||
CF135139.1 | - | 3, 4 | 402 | |||
CN313455.1 | - | 4, 5 | 522 |