Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR17HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027350.1 | 2 | 1899 | |||
GenBank mRNA | BC109081.2 | - | 2 | 1777 | ||
BC109082.2 | - | 3 | 1392 |
Gene Symbol: MIR18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AB176708.1 | - | LIT3364 oncomir-1 |
2 | 1899 |