Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145531.2 | NP_001139003.1 | 4 | 1935 | ||
NM_001302625.1 | NP_001289554.1 | 3 | 1275 | |||
XM_017012010.1 | XP_016867499.1 | 4 | 4425 | |||
XM_017012011.1 | XP_016867500.1 | 5 | 4311 | |||
XM_017012012.1 | XP_016867501.1 | 4 | 4037 | |||
XM_017012013.1 | XP_016867502.1 | 5 | 3953 | |||
XM_017012014.1 | XP_016867503.1 | 4 | 4033 | |||
GenBank mRNA | AK093321.1 | - | 4 | 1935 | ||
CA390440.1 | - | 1 | 402 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000614528.3 | - | 1907 | Chr. - - [-] on Build |