Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC087855.1 | - | 4 | 1488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000437751.1 | - | 4 | 1679 | Chr. 2 - 131402901 - 131409049 [+] on Build GRCh38 |