Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINCR-0002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120606.1 | 7 | 1084 | |||
GenBank mRNA | BX365943.2 | - | 4 | 802 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000439804.5 | - | 4 | 803 | Chr. 3 - 191557324 - 191590356 [+] on Build GRCh38 |