Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131243.1 | 1, 2 | 119 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000456090.2 | - | 1, 2 | 119 | Chr. 17 - 17674026 - 17677688 [-] on Build GRCh38 |