Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100129175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046086.1 | 3 | 866 | |||
GenBank mRNA | AK092605.1 | - | 3 | 866 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441450.1 | - | 3 | 866 | Chr. 2 - 219002215 - 219015721 [+] on Build GRCh38 |