Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047499.1 | 3 | 484 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000417473.6 | - | 3 | 484 | Chr. 2 - 11393981 - 11403077 [+] on Build GRCh38 |