Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC253573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033844.1 | 7 | 1964 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000427914.5 | - | 7 | 1964 | Chr. 3 - 186454991 - 186493661 [-] on Build GRCh38 |