Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVIL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003930.2 | 3 | 852 | |||
NR_110923.1 | 3 | 878 | ||||
NR_110924.1 | 3 | 877 | ||||
GenBank mRNA | BC031656.1 | - | 3 | 807 | ||
BC107795.1 | - | 3 | 861 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000427063.6 | - | 3 | 814 | Chr. 10 - 29409572 - 29422372 [+] on Build GRCh38 |