Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002869.1 | XP_016858358.1 | 6 | 12955 |
Gene Symbol: LOC100506459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126833.1 | - | 2 | 2269 | ||
AL832882.1 | - | 1 | 4361 |
Gene Symbol: LOC105378922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018791.1 | - | 1 | 1208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000456414.2 | - | 2 | 2269 | Chr. 1 - 116493023 - 116499212 [-] on Build GRCh38 |