Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046546.1 | 1 | 94 | |||
GenBank mRNA | BX089534.1 | - | 1 | 94 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455967.1 | - | 1 | 94 | Chr. 1 - 110208834 - 110209987 [-] on Build GRCh38 |