Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047036.1 | 2 | 112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000440160.1 | - | 2 | 112 | Chr. 13 - 34435450 - 34640685 [-] on Build GRCh38 |