Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038222.1 | 5 | 914 | |||
GenBank mRNA | AK056703.1 | - | 5 | 904 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000445498.5 | - | 5 | 904 | Chr. 10 - 11849613 - 11894700 [-] on Build GRCh38 |