Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105375304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136270.1 | 2 | 660 | |||
NR_136271.1 | 2 | 572 | ||||
NR_136272.1 | 2 | 467 | ||||
GenBank mRNA | BG208557.1 | - | 1 | 396 | ||
CK824920.1 | - | 2 | 525 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000451368.1 | - | 2 | 575 | Chr. 7 - 26372144 - 26376272 [-] on Build GRCh38 |