Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110707.1 | 2 | 241 | |||
NR_110708.1 | 2 | 241 | ||||
GenBank mRNA | BM701845.1 | - | 2 | 242 | ||
BQ049787.1 | - | 2 | 202 | |||
DA064533.1 | - | 2 | 226 | |||
DA195154.1 | - | 2 | 226 |