Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110827.1 | 6 | 1749 | |||
GenBank mRNA | BC034818.1 | - | 6 | 1749 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419257.1 | - | 6 | 1749 | Chr. 17 - 72072333 - 72093481 [+] on Build GRCh38 |