Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103743.1 | 2 | 1361 | |||
NR_103744.1 | 2 | 595 | ||||
GenBank mRNA | BC047723.1 | - | 2 | 581 | ||
BQ708590.1 | - | 2 | 458 | |||
BX648855.1 | - | 2 | 1315 |