Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046539.2 | 4 | 396 | |||
NR_051993.2 | 3 | 236 | ||||
NR_051994.2 | 4 | 324 | ||||
NR_109838.1 | 5 | 515 | ||||
GenBank mRNA | BX093461.1 | - | 4 | 395 | ||
BX398653.2 | - | 5 | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435636.5 | - | 4 | 363 | Chr. 13 - 60012751 - 60044357 [+] on Build GRCh38 |