Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ43879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034162.1 | 4 | 1632 | |||
GenBank mRNA | AK125867.1 | - | 4 | 1632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455228.1 | - | 4 | 1632 | Chr. 2 - 238919302 - 238926269 [-] on Build GRCh38 |