Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WEE2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015392.1 | 7 | 884 | |||
GenBank mRNA | AK098171.1 | - | 7 | 884 | ||
BC101358.2 | - | 4 | 328 | |||
BC101359.2 | - | 7 | 1138 | |||
BC101360.2 | - | 3 | 174 | |||
BC131564.1 | - | 5 | 514 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB044037.1 | - | 6 | 568 | Chr. 7 - 141705703 - 141738230 [-] on Build GRCh38 |