Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 1609 | |||
NR_110066.1 | 3 | 1291 | ||||
NR_110067.1 | 4 | 1547 | ||||
NR_110068.1 | 2 | 1247 | ||||
NR_110069.1 | 3 | 1439 | ||||
NR_110070.1 | 5 | 1398 | ||||
GenBank mRNA | AK090593.1 | - | 3 | 1173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000423008.1 | - | 3 | 1291 | Chr. 7 - 1160374 - 1165267 [+] on Build GRCh38 |