Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278674.1 | NP_001265603.1 | 5 | 887 | ||
GenBank mRNA | BC068105.1 | - | 3 | 435 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000426859.1 | - | 5 | 887 | Chr. 13 - 113895103 - 113921317 [+] on Build GRCh38 |