Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105378470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134329.1 | 1 | 129 | |||
NR_134330.1 | 1 | 129 | ||||
NR_134331.1 | 1 | 129 | ||||
GenBank mRNA | BC036309.1 | - | 1 | 127 | ||
BG722736.1 | - | 1 | 132 | |||
BX098586.1 | - | 1 | 98 | |||
DB026284.1 | - | 1 | 84 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000449761.5 | - | 1 | 30 | Chr. 10 - 106140264 - 106188722 [+] on Build GRCh38 |