Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102430.1 | 3 | 609 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431361.1 | - | 3 | 609 | Chr. 20 - 62800627 - 62805587 [-] on Build GRCh38 |