Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105376736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135061.1 | 2 | 479 | |||
NR_135062.1 | 2 | 383 | ||||
NR_135063.1 | 2 | 309 | ||||
GenBank mRNA | AI342698.1 | - | 2 | 383 | ||
BG184665.1 | - | 1 | 249 |