Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101926966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110201.1 | 6 | 772 | |||
GenBank mRNA | AK093525.1 | - | 6 | 772 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000492352.5 | - | 6 | 772 | Chr. 2 - 15690782 - 15718961 [+] on Build GRCh38 |