Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100133985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024444.1 | 1 | 135 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000414141.1 | - | 1 | 136 | Chr. 2 - 70124036 - 70125317 [-] on Build GRCh38 |