Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109943.1 | 3 | 665 | |||
GenBank mRNA | BX091132.1 | - | 2 | 421 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000411453.2 | - | 645 | Chr. 20 - 50172590 - 50176559 [+] on Build GRCh38 |