Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105373805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136318.1 | 3 | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429796.1 | - | 2 | 422 | Chr. 2 - 191030751 - 191032314 [+] on Build GRCh38 |