Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100507507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322263.1 | NP_001309192.1 | 2 | 1127 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429630.1 | - | 2 | 597 | Chr. 7 - 143379692 - 143380495 [-] on Build GRCh38 |