Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110570.1 | 3 | 360 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000413525.1 | - | 2 | 299 | Chr. 2 - 144566435 - 144579434 [+] on Build GRCh38 |