Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033202.2 | 3 | 286 | |||
NR_033203.1 | 3 | 189 | ||||
NR_033205.1 | 3 | 282 | ||||
NR_110329.1 | 2 | 86 | ||||
NR_110330.1 | 2 | 183 | ||||
NR_110331.1 | 4 | 522 | ||||
GenBank mRNA | AA299267.1 | - | 3 | 226 | ||
BG196429.1 | - | 2 | 91 | |||
BG203124.1 | - | 3 | 317 | |||
BG203609.1 | - | 3 | 312 | |||
BG217847.1 | - | 2 | 180 | |||
CB046509.1 | - | 3 | 198 | |||
DA741273.1 | - | 4 | 527 |