Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108099.1 | 2 | 1211 | |||
GenBank mRNA | BC035176.1 | - | 2 | 1216 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429367.1 | - | 2 | 1211 | Chr. 7 - 56534230 - 56538130 [-] on Build GRCh38 |